missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Biohazard Warning Tape
GREENER_CHOICE

Catalog No. 15965
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15965 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15965
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for taping down bench protectors to delineate hazardous areas.

  • Adhesive tape is printed with the word “BIOHAZARD” and the biohazard warning symbol
  • Prevents curling and helps prevent spills from seeping under protective matting
  • Can be cut into labels (250 labels per roll)
  • Red fluorescent color

Order Info

Roll size: 500L x 0.50 in. W (1270 x 1.3cm).

Specifications

Adhesive Type Self-adhesive
Color Red
Format Tape
Product Type Biohazard Warning Tape
For Use With (Application) Ideal for taping down bench protectors to delineate hazardous areas
Quantity Each
Length (English) 500 in.
Length (Metric) 12.7 m
Width (English) 0.5 in.