missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Adhesive Plate Seals

Catalog No. 08408240
Change view
Click to view available options
Material:
Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
08408240 Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08408240
Get Up to $2,500 in Products with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Seals are optically clear for PCR applications

  • Seals most plates
  • Rated from -40 to ±125°C
  • Polypropylene

Specifications

Color Clear
Material Polypropylene
Sterility Non-sterile
No. per Case 100
For Use With (Application) For PCR applications
Product Type Plate Seal
Temperature Range (Metric) -40°C to +125°C