Learn More
Fisherbrand™ Preparative Marker for Horizontal Midigel Systems
Catalog No. FBSB1336
Change view
Click to view available options
Teeth Count:
2
Catalog No. Teeth Count
FBSB1336 2
1 options
Catalog No. FBSB1336 Supplier Fisherbrand™ Supplier No. A1 PREP
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Preparative Marker for Horizontal Midigel Systems
  • Preparative/marker for 1.5mm comb
Specifications
For Use With (Equipment) Owl A1 Large Gel Electrophoresis System
Type Preperative Marker
Teeth Count 2
Thickness (Metric) Teeth 0.15 cm
Volume (Fill) Metric 875/35 μL
Width (English) Teeth 4.6/0.19 in.
Width (Metric) Teeth 11.7/0.5 cm