Learn More
Fisherbrand™ Vortex Mixer Accessories: Foam Racks
Catalog No. 02215387
Change view
Click to view available options
Product Type:
Microplate Holder
Microtube Foam rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
02215387 Microplate Holder
02215386 Microtube Foam rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02215387 Supplier Fisherbrand™ Supplier No. 80862998
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Fisherbrand vortex mixers.
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Mixes 96-well plates or deepwell blocks
  • Requires Insert Retainer (02-215-391)
Specifications
Product Type Microplate Holder
Material Foam
For Use With (Equipment) Digital and analog mini vortex mixer