missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Flip-Stick™ Thermometer

Multipurpose, fold-out design makes taking probe readings in any position a snap

 Afficher plus de versions de ce produit

Numéro de catalogue. 1464845


Il en reste null

Description

Description

Fisher Scientific™ Traceable™ Flip-Stick™ Thermometer with fold-out design makes taking probe readings in any position a snap.

  • Features °F/°C switch
  • Stainless-steel probe
  • Shockproof and drop-proof up to 5 ft.
  • Max/min temperature reading over any period
  • Wide temperature range cover most routine measurements
  • Position probe at any angle
  • Protect probe in fold-up position
  • 0.75 in. Display is easy to read
  • Rapid reading update—every second
  • HOLD switch freezes display to capture a temperature

Certifications

Individually serial-numbered Traceable Certificate is provided from ISO 17025 calibration laboratory accredited by A2LA indicating traceability to standards provided by NIST (National Institute of Standards and Technology).

Spécifications

Identifiants chimiques

11 x 0.6 in. (28 x 1.5 cm)
Stainless Steel probe
280 mm
0.1°C
68 g
Digital Thermometer
572°F
-58°F
-58°F to +572°F
Yes
1.5 cm
Food, gases, samples, reagents, water baths and semi solids
4.5 in.
11.4 cm
2.4 oz.
0.6 in.
11 in.
300°C
-50°C
-50°C to +300°C
Digital

Spécifications

0.14 in.
Food, gases, samples, reagents, water baths and semi solids
4.5 in.
11.4 cm
0.1°C
68 g
Digital Thermometer
572°F
-58°F
-58°F to +572°F
Yes
Digital
11 x 0.6 in. (28 x 1.5 cm)
Stainless Steel probe
280 mm
Stainless Steel
2.4 oz.
0.6 in.
11 in.
300°C
-50°C
-50°C to +300°C
±1°C
1.5 cm
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions