missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Reverse Break Casters

Numéro de catalogue. 14359001
Change view
Click to view available options
:
Reverse Brake Casters; 5/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
14359001 Reverse Brake Casters; 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 14359001
Il en reste null

Fisherbrand™ Reverse Break Casters lock when weight is applied. Rubberized coating prevents scuffing on hard floors.

  • Rubberized coating prevents scuffing on hard floors
  • Locks when weight is applied

Order Info

Sold in a pack of (5) five

Spécifications

Adjustable Yes
Color Black
For Use With (Equipment) Seating
Warranty 15 year parts, lifetime on pneumatic lift, 5 year on upholstery