Learn More
Fisherbrand™ Precision Specific Gravity 325 - 368mm Long Hydrometers
Requires 275mL liquid volume for submersion.
$54.44 - $809.38
Specifications
| Increments | 0.0005 |
|---|---|
| For Use With (Equipment) | 350mL Borosilicate Glass cylinder |
| Catalog Number | Certifications/Compliance | Length (Metric) | Specific Gravity Range | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Certifications/Compliance | Length (Metric) | Specific Gravity Range | Price | Quantity & Availability | ||||
|
11555DC
|
3-point Cert, NIST, ASTM E100, E126 | 345 mm | 0.820 to 0.890 |
Each for $729.67
|
|
||||
|
11555CC
|
3-point Cert, NIST, ASTM E100, E126 | 330 mm | 0.760 to 0.830 |
Each for $809.38
|
|
||||
|
11555EC
|
3-point Cert, NIST, ASTM E100, E126 | 340 mm | 0.880 to 0.950 |
Each for $730.07
|
|
||||
|
11555GC
|
3-point Cert, NIST, ASTM E100, E126 | 340 mm | 1.000 to 1.070 |
Each for $730.18
|
|
||||
|
11555FC
|
3-point Cert, NIST, ASTM E100, E126 | 325 mm | 0.940 to 1.010 |
Each for $730.07
|
|
||||
|
11555HC
|
3-point Cert, NIST, ASTM E100, E126 | 330 mm | 1.060 to 1.130 |
Each for $730.29
|
|
||||
|
11555JC
|
3-point Cert, NIST, ASTM E100, E126 | 335 mm | 1.120 to 1.190 |
Each for $730.29
|
|
||||
|
11555KC
|
3-point Cert, NIST, ASTM E100, E126 | 340 mm | 1.180 to 1.250 |
Each for $730.76
|
|
||||
|
11555LC
|
3-point Cert, NIST, ASTM E100, E126 | 340 mm | 1.240 to 1.310 |
Each for $730.76
|
|
||||
|
11555MC
|
3-point Cert, NIST, ASTM E100, E126 | 340 mm | 1.300 to 1.370 |
Each for $730.87
|
|
||||
|
11555U
|
NIST, ASTM E100, ASTM E126 | 330 mm | 1.660 to 1.730 |
Each for $56.76
|
|
||||
|
11555W
|
NIST, ASTM E100, ASTM E126 | 330 mm | 1.84 to 1.92 |
Each for $59.93
|
|
||||
|
11555B
|
NIST, ASTM E100, ASTM E126 | 340 mm | 0.700 to 0.770 |
Each for $54.44
|
|
||||
|
11555C
|
NIST, ASTM E100, ASTM E126 | 330 mm | 0.760 to 0.830 |
Each for $54.44
|
|
||||
|
11555E
|
NIST, ASTM E100, ASTM E126 | 340 mm | 0.880 to 0.950 |
Each for $54.44
|
|
||||
|
11555F
|
NIST, ASTM E100, ASTM E126 | 325 mm | 0.940 to 1.010 |
Each for $54.44
|
|
||||
|
11555D
|
NIST, ASTM E100, ASTM E126 | 345 mm | 0.820 to 0.890 |
Each for $54.44
|
|
||||
|
11555J
|
NIST, ASTM E100, ASTM E126 | 335 mm | 1.120 to 1.190 |
Each for $54.44
|
|
||||
|
11555H
|
NIST, ASTM E100, ASTM E126 | 330 mm | 1.060 to 1.130 |
Each for $54.44
|
|
||||
|
11555K
|
NIST, ASTM E100, ASTM E126 | 340 mm | 1.180 to 1.250 |
Each for $54.44
|
|
||||
|
11555L
|
NIST, ASTM E100, ASTM E126 | 340 mm | 1.240 to 1.310 |
Each for $54.44
|
|
||||
|
11555P
|
NIST, ASTM E100, ASTM E126 | 345 mm | 1.420 to 1.490 |
Each for $54.44
|
|
||||
|
11555M
|
NIST, ASTM E100, ASTM E126 | 340 mm | 1.300 to 1.370 |
Each for $54.44
|
|
||||
|
11555Q
|
NIST, ASTM E100, ASTM E126 | 330 mm | 1.48 to 1.55 |
Each for $54.87
|
|
||||
|
11555N
|
NIST, ASTM E100, ASTM E126 | 345 mm | 1.360 to 1.430 |
Each for $54.44
|
|
||||
|
11555R
|
NIST, ASTM E100, ASTM E126 | 330 mm | 1.54 to 1.61 |
Each for $54.55
|
|
||||
|
11555S
|
NIST, ASTM E100, ASTM E126 | 330 mm | 1.6 to 1.67 |
Each for $54.44
|
|
||||
|
11555T
|
NIST, ASTM E100, ASTM E126 | 330 mm | 1.78 to 1.85 |
Each for $54.65
|
|
||||
|
11555A
|
NIST, ASTM E100, ASTM E126 | 340 mm | 0.640 to 0.710 |
Each for $54.44
|
|
||||
|
11555G
|
NIST, ASTM E100, ASTM E126 | 340 mm | 1.000 to 1.070 |
Each for $54.44
|
|
||||
Description
Requires 275mL liquid volume for submersion.
- Use Cylinder 08-530K, available separately
- Permanently marked with individual serial numbers
- Accuracy meets or exceeds ASTM E 100 specifications
- Calibrated according to ASTM E 126 specifications
- With divisions of 0.001
- Use Cylinder 08-530E, available separately
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™.
Specifications
| 0.0005 | |
| 350mL Borosilicate Glass cylinder |