missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Horizontal Mini Gel Electrophoresis System

Catalog No. 14955170
Change view
Click to view available options
Gel Dimensions:
10.5 x 6 cm or 5 x 6 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gel Dimensions
14955170 10.5 x 6 cm or 5 x 6 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14955170 Supplier Fisherbrand™ Supplier No. E2102F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for quick separation of DNA or nucleic acids in agarose mini-gels, the Fisherbrand Mini Horizontal Gel System is a complete, compact mini gel electrophoresis system for easy nucleic acid separation. The horizontal system comes with an attached power supply, two casting stands, six gel trays and four combs — everything necessary for DNA or nucleic acid, horizontal electrophoresis.

Complete system - no additional components required

Order Info

Complete system - no additional components required

†Compatible with multi-channel pipettes.

Specifications

Description Horizontal Mini Gel Electrophoresis System with integrated power supply
Timer 0 to 99 min. or continuous
Type Horizontal Gel Electrophoresis System
Display Digital
Gel Dimensions 10.5 x 6 cm or 5 x 6 cm
Includes Gel tank, power supply, two casting stands, two 10.5 x 6cm gel trays and 2 dual format combs (12/22 teeth), four 5 x 6cm gel and 2 dual format combs (18/10 teeth)
No. of Gels One 10.5 x 6 cm or two 5 x 6 cm
Quantity 1 Each
Volume (English) Buffer 10 oz.
Volume (Metric) Buffer 300 mL
Warranty 2 Years
Dimensions (L x W x H) 19 x 13 x 5.5 cm (7.5 x 5.1 x 2.1 in.)
Length (English) 5.1 in.
Length (English) Gel 2.36 in.
Length (Metric) 13 cm
Length (Metric) Gel 6 cm
Width (English) 7.5 in.
Width (English) Gel 4.1 in. or 2.4 in.
Width (Metric) 19 cm
Width (Metric) Gel 10.5 cm or 5 cm
Height (English) 2.1 in.
Height (Metric) 5.5 cm
Weight (English) 1 lb.
Weight (Metric) 0.45 kg
Electrical Requirements 115 V 50/60 Hz
Voltage 35, 50 or 100 V (output)
Show More Show Less