missing translation for 'onlineSavingsMsg'
Learn More

Fisher Science Education™ Balloons

Catalog No. S04187
Change view
Click to view available options
Color:
Assorted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S04187 Assorted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04187 Supplier Fisher Science Education™ Supplier No. BL5200144PK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Vibrant, assorted color latex balloons can be filled with air or helium.

  • Diameter: 9 in.
  • Assorted Colors

Specifications

Product Type Balloon
Color Assorted
For Use With (Application) For science classroom or with kits
Shape Round
Diameter (English) Outer 9 in.
Diameter (Metric) Outer 23 cm