missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Proteinase K (Tritirachium album/Molecular Biology)
$191.68 - $1,150.53
Specifications
| Color | White |
|---|---|
| For Use With (Application) | Isolation of DNA and RNA from cell and tissue preparations, inactivating many Enzymes, including Endogenous Nucleases |
| Content And Storage | -20°C |
| Source | Tritirachium album |
| Product Type | Proteinase K |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
BP170050
|
Fisher BioReagents™
BP1700 50 |
50 mg |
Each for $191.68
|
|
|||||
|
BP1700100
|
Fisher BioReagents™
BP1700 100 |
100 mg |
Each for $255.29
|
|
|||||
|
BP1700500
|
Fisher BioReagents™
BP1700 500 |
500 mg |
Each for $1,150.53
|
|
|||||
Description
Proteinase K (Tritirachium album/Molecular Biology)
- Not on TSCA inventory: for research and development use only; not for manufacturing or commercial purposes
Proteinase K is widely used in the isolation of DNA and RNA from cell and tissue preparations. It is a wide-spectrum protease, inactivating many enzymes, including endogenous nucleases.
Specifications
| White | |
| -20°C | |
| Proteinase K |
| Isolation of DNA and RNA from cell and tissue preparations, inactivating many Enzymes, including Endogenous Nucleases | |
| Tritirachium album | |
| Solid |