missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Proteinase K (Tritirachium album/Molecular Biology)

Catalog No. BP1700500
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP1700500 500 mg
BP170050 50 mg
BP1700100 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP1700500 Supplier Fisher BioReagents™ Supplier No. BP1700 500
Only null left

Proteinase K (Tritirachium album/Molecular Biology)

  • Not on TSCA inventory: for research and development use only; not for manufacturing or commercial purposes

Proteinase K is widely used in the isolation of DNA and RNA from cell and tissue preparations. It is a wide-spectrum protease, inactivating many enzymes, including endogenous nucleases.

Specifications

Color White
For Use With (Application) Isolation of DNA and RNA from cell and tissue preparations, inactivating many Enzymes, including Endogenous Nucleases
Content And Storage -20°C
Source Tritirachium album
Quantity 500 mg
Product Type Proteinase K
Form Solid