Learn More
Thermo Scientific™ Finntip™ Stepper Pipette Tips
GREENER_CHOICE
Integrate Thermo ScientificTM FinntipTM Stepper Pipette Tips with the Thermo ScientificTM FinnpipetteTM Stepper Pipetter.
Supplier:  Thermo Scientific™ 9404213
 View more versions of this product
Catalog No. 21377157
Only null left
Description
Description
Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.
  • Nonsterile tips are bulk packaged; sterile tips are individually wrapped
Specifications
Chemical Identifiers
Non-filtered
ISO 9001, ISO 14001
Thermo Scientific FinnPipette Stepper
FinnTip
Specifications
12.5 mL
Non-filtered
ISO 9001, ISO 14001
Thermo Scientific FinnPipette Stepper
250, 500, 750, 1000, 1250 μL
Sterile
Individually Wrapped
FinnTip
Product Suggestions
Product Suggestions