Learn More
Thermo Scientific™ Finnpipette™ Stands
GREENER_CHOICE
Catalog No. 1438714
Change view
Click to view available options
Product Type:
Carousel Stand with Cover
Pipette Stand
Holds:
Electronic Pipette
Manual Pipettes (3)
Manual Pipettes (6)
Manual Pipettes (6) or Electronic Pipettes (3)
Multi-channel Manual Pipette
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Holds
1438714 Pipette Stand Manual Pipettes (6)
14245160 Pipette Stand Manual Pipettes (6)
1438715 Pipette Stand Manual Pipettes (3)
21377002 Carousel Stand with Cover Manual Pipettes (6) or Electronic Pipettes (3)
1438721 Pipette Stand Electronic Pipette
14245161 Pipette Stand Multi-channel Manual Pipette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 1438714 Supplier Thermo Scientific™ Supplier No. 9420290
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Provide safe and convenient storage for manual and electronic pipettes using these stands, with options for single stand, F-stand, multi-stand, multichannel and carousel.
Provide safe and convenient storage for manual and electronic pipettes using Thermo Scientific™ Finnpipette™ Stands. Available options include options for single stand, F-stand, multi-stand, multichannel and carousel.
Finnpipette F-Stand
  • Accommodates six pipettes
  • Sturdy crossbar structure with slot for each pipette for secure storage
  • Suitable for both single channel and multichannel models
Finnpipette Multichannel Stand
  • Multi-position stand
  • Can hold any multichannel Finnpipette
  • Pipette can be stored in upright or horizontal position
  • Small footprint
Finnpipette Carousel Stand
  • Rotating, portable
  • Transparent lid protects pipettes from contamination
  • Shipped with three adapters for electronic Finnpipette Novus pipettes and five adapters for manual Finnpipette pipettes
Finnpipette Novus Stand
  • For convenient storage of electronic Finnpipette Novus pipette
  • Accommodates both single and multichannel Finnpipette Novus models with ease
Specifications
Color Gray
Holds Manual Pipettes (6)
Positions 6
Product Type Pipette Stand