missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finnpipette™ F2 Good Laboratory Pipetting (GLP) Kits
GREENER_CHOICE

Catalog No. 05719621
Change view
Click to view available options
Volume (Metric):
0.2 to 1000 μL
1 to 1000 μL
10 to 10,000 μL
2 to 1000 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
05719621 2 to 1000 μL
05719510 1 to 1000 μL
05719511 0.2 to 1000 μL
05719512 10 to 10,000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 05719621 Supplier Thermo Scientific™ Supplier No. 4701070
Get Three Cases of Pipette Tips with Kit Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enjoy the convenience of having a variety of Finnpipette™ F2 single channels packaged together, in a range of volumes, with compatible Finntip pipette tips and a pipette stand to support good laboratory pipetting in your lab

Access everything needed for pipetting in these all-inclusive kits. Thermo Scientific™ Finnpipette™ F2 GLP (Good Laboratory Pipetting) Kits provide durable, high-performance Finnpipette F2 pipettes which offer maximum reliability and comfort. Built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light, these variable volume pipettes are fully autoclavable without disassembling.

Features:

  • AVG-volume gearing mechanism for accuracy and precision
  • Fully autoclavable for protection
  • Soft-touch tip ejector for light tip ejection
  • Large display for better vision ergonomics
  • Very light pipetting forces

Kit 1 includes:

  • Three Thermo Scientific Finnpipette Pipetters from 1 to 1000 μL:
    • Finnpipette F2
    • 1-10 μL
    • 10-100 μL
    • 100-1000 μL
  • Flex tips: 10, 1 x 96; 200, 1 x 96; 1000, 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 2 includes:

  • Four Thermo Scientific Finnpipette Pipetters from 0.2 to 1000 μL:
    • Finnpipette F2
    • 0.2-2 μL
    • 2-20 μL univ
    • 20-200 μL
    • 100-1000 μL
  • Flex tips: 10, 12 x 96; 200, 2 x 96; 1000, 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 3 includes:

  • Three Thermo Scientific Finnpipette Pipetters from 10 to 10,000 μL:
    • Finnpipette F2
    • 10-100 μL, 100-1000 μL
    • 1000-10,000μL
  • Flex tips: 200, 1 x 96; 1000, 1 x 96; FT 10mL, 1 x 24 F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 4 includes:

  • Three Thermo Scientific Finnpipette Pipetters from 20 to 1000μL:
    • Finnpipette F2
    • 2-20μL, 20-200μL
    • 100-1000 μL 
  • Flex tips: (2) 200 μL, 10 x 96; 100-1000 μL, 10 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Ordering Information:

The kit contains samples of Finntip Flex, the most advanced traditional tip offering very low attachment and ejection forces. The Good Laboratory Pipetting Guide contains all the information of pipetting from choosing the right type to detailed de-contamination information.

Warranty: 5 years with web registration.

Specifications

Compatible Tips Flex Tips: 200, 1000 μL
Description Finnpipette F2 GLP Kit 4: 2-20μL, 20-200μL and 100-1000μL
Autoclavable Autoclavable
Color Assorted
Model F2
Product Line Finnpipette F2
Type GLP Kit
Volume (Metric) 2 to 1000 μL