missing translation for 'onlineSavingsMsg'
Learn More

Lynn Peavey Fingerprint Ink

Numéro de catalogue. p-2829231
Change view
Click to view available options
Weight (English):
2 oz.
4 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Weight (English)
S69019A 2 oz.
S69019B 4 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. S69019A Fournisseur Lynn Peavey Code fournisseur 5569
Il en reste null

Quick drying black ink produces excellent ridge detail.

Spécifications

Color Black
For Use With (Application) Provides excellent ridge detail
Weight (English) 2 oz.
Product Type Fingerprint Ink