missing translation for 'onlineSavingsMsg'
Learn More

Lynn Peavey Fingerprint Ink

Catalog No. S69019B
Change view
Click to view available options
Weight (English):
2 oz.
4 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
S69019B 4 oz.
S69019A 2 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S69019B Supplier Lynn Peavey Supplier No. 5570

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Quick drying black ink produces excellent ridge detail.

Specifications

Color Black
For Use With (Application) Provides excellent ridge detail
Weight (English) 4 oz.
Product Type Fingerprint Ink