World Precision Mini Iris Scissors, 8cm, Straight, Sharp Tips
Numéro de catalogue. NC1564460
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Numéro de catalogue.
NC1564460
Fournisseur
World PrecisionCode fournisseur
503670
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.