missing translation for 'onlineSavingsMsg'
Learn More

Finasteride 98.0+%, TCI America™

Catalog No. F0675200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0675200MG 200 mg
F06751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0675200MG Supplier TCI America Supplier No. F0675200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98319-26-7
Molecular Formula C23H36N2O2
Molecular Weight (g/mol) 372.553
MDL Number MFCD00869737
InChI Key DBEPLOCGEIEOCV-WSBQPABSSA-N
Synonym finasteride, proscar, propecia, finastid, prostide, chibro-proscar, finasterida, finasteridum, finpecia, finasteridum inn-latin
PubChem CID 57363
ChEBI CHEBI:5062
IUPAC Name (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide
SMILES CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C

Specifications

Melting Point 253°C
Color White
Quantity 200 mg
Formula Weight 372.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Finasteride
RTECSNumber CL5245000
TSCA No