Learn More
Alkali Scientific Film Sealing Paddle, 1 bag of 50
Numéro de catalogue. 502389419
Encompass
The Multi-Swipe Film Sealing Paddle ensures a secure and uniform, pressure-sensitive seal around all wells and is an economical alternative to plate rollers. Film-Sealing Paddle are designed for use with all plate types. Reusable and recyclable. The semi-soft, pliable sealing surfaces prevent the film/foil from being torn or ripped. Features two unique sized corner radius: 1/8 inch (0.3175 centimeter) and 1/16 inch (0.1588 centimeter) to seal around the plate perimeter; the tighter radii is for use with raised-rim PCR plates. Features two sealing surfaces: the shorter length fits inside the walls of raised-rim plates; the longer length is ideal for flat-top and deep-well plates. The non-flat contoured side is designed to go down to the plate; the center raised textured surface provides a non-slip finger hold.
Numéro de catalogue. 502389419 Fournisseur Alkali Scientific Code fournisseur SF15ML
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The Multi-Swipe Film Sealing Paddle ensures a secure and uniform, pressure-sensitive seal around all wells and is an economical alternative to plate rollers. Film-Sealing Paddle are designed for use with all plate types. Reusable and recyclable. The semi-soft, pliable sealing surfaces prevent the film/foil from being torn or ripped. Features two unique sized corner radius: 1/8 inch (0.3175 centimeter) and 1/16 inch (0.1588 centimeter) to seal around the plate perimeter; the tighter radii is for use with raised-rim PCR plates. Features two sealing surfaces: the shorter length fits inside the walls of raised-rim plates; the longer length is ideal for flat-top and deep-well plates. The non-flat contoured side is designed to go down to the plate; the center raised textured surface provides a non-slip finger hold.