Learn More
BRANDTECH™ BRAND™ Filling Tubes for BRAND™ Bottletop Dispensers and Burettes
Replacement filling tubes for BRAND™ bottletop instruments, including Dispensette™ S, Dispensette™ S Organic, and Dispensette™ S Trace Analysis dispensers; seripettor™ and seripettor™ pro dispensers; and Titrette™ bottletop burettes.
$103.71 - $218.82
| Catalog Number | Mfr. No. | Length (Metric) | Sterility | For Use With (Equipment) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (Metric) | Sterility | For Use With (Equipment) | Price | Quantity | |||
|
14-380-911
|
BRANDTECH™ BRAND™
708210 |
70 to 140 mm | Non-sterile | Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL) |
Each for $103.71
|
|
|||
|
14-380-912
|
BRANDTECH™ BRAND™
708212 |
125 to 240 mm | Non-sterile | Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL) |
Each for $143.88
|
|
|||
|
14-380-913
|
BRANDTECH™ BRAND™
708214 |
195 to 350 mm | Non-sterile | Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL) |
Each for $205.03
|
|
|||
|
14-380-914
|
BRANDTECH™ BRAND™
708216 |
250 to 480 mm | Non-sterile | Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL) |
Each for $163.68
|
|
|||
|
14-380-915
|
BRANDTECH™ BRAND™
708218 |
170 to 330 mm | Non-sterile | Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (25, 50, 100 mL), Seripettor™ pro (25 mL), Titrette™ (10, 25, 50 mL) |
Each for $208.65
|
|
|||
|
14-380-916
|
BRANDTECH™ BRAND™
708220 |
250 to 480 mm | Non-sterile | Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (25, 50, 100 mL), Seripettor™ pro (25 mL), Titrette™ (10, 25, 50 mL) |
Each for $218.82
|
|
|||
Description
- Telescoping FEP filling tubes adjust to bottle height without cutting
- Compatible with Dispensette™ S, Dispensette™ S Organic, Dispensette™ S Trace Analysis; seripettor™ pro; and Titrette™
- Also compatible with Dispensette™ III
- Standard PP filling tubes can be manually cut to fit various bottle sizes
- Compatible only with seripettor™
- Available sterile with O-ring
- Available in multiple lengths based on dispenser nominal volume
- Design supports aspiration from the bottom of the bottle while reducing precipitate intake
Specifications
| Clear |