missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Filling Tubes for BRAND™ Bottletop Dispensers and Burettes

Catalog No. 14380916
Change view
Click to view available options
Length (Metric):
125 to 240 mm
170 to 330 mm
195 to 350 mm
250 to 480 mm
70 to 140 mm
Sterility:
Non-sterile
For Use With (Equipment):
Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL)
Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (25, 50, 100 mL), Seripettor™ pro (25 mL), Titrette™ (10, 25, 50 mL)
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Sterility For Use With (Equipment)
14380916 250 to 480 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (25, 50, 100 mL), Seripettor™ pro (25 mL), Titrette™ (10, 25, 50 mL)
14380911 70 to 140 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL)
14380912 125 to 240 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL)
14380913 195 to 350 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL)
14380914 250 to 480 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL)
14380915 170 to 330 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (25, 50, 100 mL), Seripettor™ pro (25 mL), Titrette™ (10, 25, 50 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14380916 Supplier BRANDTECH™ BRAND™ Supplier No. 708220

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Replacement filling tubes for BRAND™ bottletop instruments, including Dispensette™ S, Dispensette™ S Organic, and Dispensette™ S Trace Analysis dispensers; seripettor™ and seripettor™ pro dispensers; and Titrette™ bottletop burettes.

  • Telescoping FEP filling tubes adjust to bottle height without cutting
    • Compatible with Dispensette™ S, Dispensette™ S Organic, Dispensette™ S Trace Analysis; seripettor™ pro; and Titrette™
    • Also compatible with Dispensette™ III
  • Standard PP filling tubes can be manually cut to fit various bottle sizes
    • Compatible only with seripettor™
    • Available sterile with O-ring
  • Available in multiple lengths based on dispenser nominal volume
  • Design supports aspiration from the bottom of the bottle while reducing precipitate intake

Specifications

Color Clear
Material FEP
For Use With (Equipment) Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (25, 50, 100 mL), Seripettor™ pro (25 mL), Titrette™ (10, 25, 50 mL)
Sterility Non-sterile
Length (Metric) 250 to 480 mm