Learn More
BRANDTECH™ BRAND™ Filling Tubes for BRAND™ Bottletop Dispensers and Burettes
Replacement filling tubes for BRAND™ bottletop instruments, including Dispensette™ S, Dispensette™ S Organic, and Dispensette™ S Trace Analysis dispensers; seripettor™ and seripettor™ pro dispensers; and Titrette™ bottletop burettes.
Supplier:  BRANDTECH™ BRAND™ 708214
 View more versions of this product
Catalog No. 14-380-913
Only null left
Explore available promotions
Description
Description
  • Telescoping FEP filling tubes adjust to bottle height without cutting
    • Compatible with Dispensette™ S, Dispensette™ S Organic, Dispensette™ S Trace Analysis; seripettor™ pro; and Titrette™
    • Also compatible with Dispensette™ III
  • Standard PP filling tubes can be manually cut to fit various bottle sizes
    • Compatible only with seripettor™
    • Available sterile with O-ring
  • Available in multiple lengths based on dispenser nominal volume
  • Design supports aspiration from the bottom of the bottle while reducing precipitate intake
Specifications
Specifications
Clear
Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL)
195 to 350 mm
FEP
Non-sterile
Product Suggestions
Product Suggestions