missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fibrinogen Beta Chain Monoclonal Antibody (2#)
GREENER_CHOICE

Numéro de catalogue. PIMA541586
Modifier l'affichage
Click to view available options
Quantité:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PIMA541586 200 μL
PIMA543823 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. PIMA541586 Fournisseur Invitrogen™ Code fournisseur MA541586

Mouse Monoclonal Antibody

Fibrinogen Beta Chain Monoclonal Antibody for Western Blot, IHC (P)

FIBB is the beta component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in its gene lead to several disorders, including afibrinogenemia, dysfibrinogenemia, hypodysfibrinogenemia and thrombotic tendency.

Spécifications

Antigène Fibrinogen Beta Chain
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2#
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gène Fgb
Numéro d’acquisition du gène P02675
Alias du gène 2510049G14Rik; Ab1-181; Ab1-216; Ac1-581; B beta polypeptide; beta-fibrinogen; epididymis secretory sperm binding protein Li 78p; Fgb; fibrinogen; Fibrinogen beta chain; fibrinogen, B beta polypeptide; fibrinogen, beta polypeptide; Fibrinopeptide B; HEL-S-78p; Liver regeneration-related protein LRRG036/LRRG043/LRRG189; MGC104327; MGC120405
Symboles de gène Fgb
Espèces hôtes Mouse
Immunogène Recombinant protein Fibrinogen Beta Chain. The antigen corresponds to amino acid range 45-491 of the target protein.
Méthode de purification Protein A/G
Quantité 200 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 2244
Espèces cibles Human
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG2b κ
Afficher plus de résultats Afficher moins de résultats