missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Fiberglass Danger Sign: FLAMMABLE MATERIALS

Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

$26.07 - $43.64

Spécifications

Finish Not Laminated
Material Fiberglass
Quantity 1 Ea.
Adhesive Type Non-adhesive
Mounting Type Corner Holes
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Dimensions (L x W) Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Dimensions (L x W) Prix Quantity & Availability  
19106206
Afficher les documents Détails de la promotion
Brady™
131793
10 x 7 in.
chaque for $26.07
Il en reste null
 
19106207
Afficher les documents Détails de la promotion
Brady™
131794
14 x 10 in.
chaque for $43.64
Il en reste null
 
Description

Description

  • Fiberglass material is abrasion-resistant and chemical-resistant
  • Fiberglass out-performs all other sign materials in terms of strength, durability, and longevity
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • For indoor and outdoor use
  • Fiberglass out-performs all other sign materials in terms of strength, durability, and longevity
Spécifications

Spécifications

Not Laminated
1 Ea.
Corner Holes
-40°F
Rectangular
FLAMMABLE MATERIALS
Fiberglass
Non-adhesive
194°F
0.0984 in.
-40°F to 194°F
Red on White
Videos
SDS
Documents

Documents