missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals FerroZine™ Iron Reagent, Hydrate, 95+%, Pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >300.0°C |
| Color | Yellow |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Assay Percent Range | 95+% |
| Solubility Information | Solubility in water: soluble |
| Formula Weight | 492.46 |
| Percent Purity | ≥95% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
RUO – Research Use Only