accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Buffers and Standards
General Chemistry Solutions
31501
Print
Ferrous Ammonium Sulfate, 0.250 N (N/4), 0.250 M (M/4), Ricca Chemical
Supplier:
Ricca Chemical Company
31501
Catalog No.
3150-1
Sign In
or
Register
to check your price and availability.
$378.50
/
Each
Quantity
Only null left
Explore available promotions
Add to cart
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Promotions
Description
Description
Specifications
Specifications
Chemical Name or Material
Ferrous Ammonium Sulfate
Grade
Laboratory
Quantity
4 L
CAS
7732-18-5
Packaging
Amber Glass Bottle
Chemical Identifiers
CAS
7732-18-5
Product Suggestions
Product Suggestions
Videos
SDS
SDS
Documents
Documents
Promotions
Promotions