missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate, 0.400 N, Ricca Chemical

Catalog No. 31521SC
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31521SC 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31521SC Supplier Ricca Chemical Company Supplier No. 31521SC

Chemical Identifiers

CAS 7783-85-9 , 7732-18-5 , 7664-93-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate
PubChem CID 115148
IUPAC Name λ2-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Green
CAS Min % 12.69, 81.0, 4.46
CAS Max % 13.2, 84.0, 4.65
Grade Laboratory
Packaging Safety-coated Glass Bottle
Quantity 4 L
Physical Form Liquid
Concentration or Composition Notes 0.400Normal
Chemical Name or Material Ferrous Ammonium Sulfate