missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate, 0.250 N (N/4), 0.250 M (M/4), Ricca Chemical

Catalog No. 315016
Change view
Click to view available options
Package: Amber Glass; Size: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
315016 Package: Amber Glass; Size: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 315016 Supplier Ricca Chemical Company Supplier No. 315016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 7783-85-9, 7732-18-5, 7664-93-9
CAS Min % 8.48, 86.0, 3.18
CAS Max % 8.82, 90.0, 3.31
Molecular Formula FeH20N2O14S2
Synonym Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate
InChI Key SHHFGSWBFZVUNE-UHFFFAOYSA-L
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name iron(3+) diammonium hexahydrate disulfate
Molecular Weight (g/mol) 392.12
PubChem CID 115148  
Packaging Amber Glass Bottle
Quantity 500 mL
Concentration or Composition Notes 0.250Normal,0.250Molar
Chemical Name or Material Ferrous Ammonium Sulfate
Show More Show Less