missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Ammonium Sulfate, 0.0250 N (N/40), 0.0250 M (M/40), Ricca Chemical
Chemical Identifiers
| CAS | 7783-85-9 , 7732-18-5 , 7664-93-9 |
|---|---|
| Molecular Formula | FeH20N2O14S2 |
| Molecular Weight (g/mol) | 392.12 |
| InChI Key | SHHFGSWBFZVUNE-UHFFFAOYSA-L |
| Synonym | Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate |
| PubChem CID | 115148 |
| IUPAC Name | iron(3+) diammonium hexahydrate disulfate |
| SMILES | [NH4+].[NH4+].O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
314216
|
N/A | 500 mL | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
314232
|
N/A | 1 L | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7783-85-9 , 7732-18-5 , 7664-93-9 | |
| 392.12 | |
| Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate | |
| iron(3+) diammonium hexahydrate disulfate |
| FeH20N2O14S2 | |
| SHHFGSWBFZVUNE-UHFFFAOYSA-L | |
| 115148 | |
| [NH4+].[NH4+].O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Specifications
| 7783-85-9 , 7732-18-5 , 7664-93-9 | |
| 0.96, 97.0, 3.59 | |
| Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate | |
| [NH4+].[NH4+].O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
| 392.12 | |
| 0.0250Normal,0.0250Molar |
| 0.92, 94.0, 3.45 | |
| FeH20N2O14S2 | |
| SHHFGSWBFZVUNE-UHFFFAOYSA-L | |
| iron(3+) diammonium hexahydrate disulfate | |
| 115148 | |
| Ferrous Ammonium Sulfate |