missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Ammonium Sulfate, 0.0250 N (N/40), 0.0250 M (M/40), Ricca Chemical
Supplier: Ricca Chemical Company 314216
| Quantity | 500 mL |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 7783-85-9 , 7732-18-5 , 7664-93-9 | |
| 392.12 | |
| Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate | |
| iron(3+) diammonium hexahydrate disulfate |
| FeH20N2O14S2 | |
| SHHFGSWBFZVUNE-UHFFFAOYSA-L | |
| 115148 | |
| [NH4+].[NH4+].O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Specifications
| Ferrous Ammonium Sulfate | |
| 0.92, 94.0, 3.45 | |
| FeH20N2O14S2 | |
| SHHFGSWBFZVUNE-UHFFFAOYSA-L | |
| iron(3+) diammonium hexahydrate disulfate | |
| 115148 | |
| 500 mL |