missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate, 0.0250 N (N/40), 0.0250 M (M/40), Ricca Chemical

Supplier:  Ricca Chemical Company 314232

Quantity 1 L
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. 314232

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

7783-85-9 , 7732-18-5 , 7664-93-9
392.12
Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate
iron(3+) diammonium hexahydrate disulfate
FeH20N2O14S2
SHHFGSWBFZVUNE-UHFFFAOYSA-L
115148  
[NH4+].[NH4+].O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Ferrous Ammonium Sulfate
0.92, 94.0, 3.45
FeH20N2O14S2
SHHFGSWBFZVUNE-UHFFFAOYSA-L
iron(3+) diammonium hexahydrate disulfate
115148  
1 L
7783-85-9 , 7732-18-5 , 7664-93-9
0.96, 97.0, 3.59
Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate
[NH4+].[NH4+].O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
392.12
Amber Glass Bottle
0.0250Normal,0.0250Molar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions