missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferroin Oxidation-Reduction Indicator, Certified, LabChem™
$84.64 - $257.39
Specifications
| Chemical Name or Material | Ferroin Oxidation-Reduction Indicator |
|---|---|
| CAS Max % | 97.81,1.49,0.7 |
| CAS | 5144-89-8 , 7782-63-0 |
| Linear Formula | FeSO4·7H2O |
| Solubility Information | Soluble in water |