missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferroin Oxidation-Reduction Indicator, Certified, LabChem™
Supplier: LabChem LC144507
Specifications
| Ferroin Oxidation-Reduction Indicator | |
| 5144-89-8,7782-63-0 | |
| Soluble in water | |
| 278.01 | |
| Amber Glass | |
| Passes Test | |
| 1g/mL | |
| 125 mL |
| 97.81,1.49,0.7 | |
| FeSO4·7H2O | |
| Passes Test | |
| Certified | |
| 1g/mL | |
| Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds | |
| Red | |
| Liquid |