missing translation for 'onlineSavingsMsg'
Learn More

Ferroin Oxidation-Reduction Indicator, Certified, LabChem™

Supplier:  LabChem LC144501

 View more versions of this product

Catalog No. LC144501

Only null left

Description

Description

Specifications

Specifications

Ferroin Oxidation-Reduction Indicator
5144-89-8 , 7782-63-0
Soluble in water
278.01
Amber Glass
Passes Test
1g/mL
500 mL
97.81,1.49,0.7
FeSO4·7H2O
Passes Test
Certified
1g/mL
Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Red
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions