missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Ferrocenylmethyl)trimethylammonium Bromide 95.0+%, TCI America™
Spécifications
Spécifications
| Quantity | 5 g |
| Formula Weight | 338.07 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (Ferrocenylmethyl)trimethylammonium Bromide |
Sécurité et manipulation
| TSCA | No |