missing translation for 'onlineSavingsMsg'
Learn More

Ferric Oxide, Red, Technical (Iron (III) Oxide), Reagents

Ferric Oxide, Red, Technical (Iron (III) Oxide), Technical, for general laboratory use. Manufactured in ISO 9001 facility.

$133.10 - $1502.13

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
Synonym Iron(III) Oxide, Rust
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-337-246
View Documents
Labchem Inc
C2205000100C2
100 g
Each for $133.10
Only null left
 
01-337-245
View Documents
Labchem Inc
C220500025KG
2.5 kg
Each for $1,502.13
Only null left
 
01-337-244
View Documents
Labchem Inc
C2205000500A5
500 g
Each for $308.23
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1309-37-1
159.69
JEIPFZHSYJVQDO-UHFFFAOYSA-N
trioxodiiron
Fe2O3
MFCD00011008
Iron(III) Oxide, Rust
[Fe][Fe](=O)(=O)=O

Specifications

3414°C
Orange/Red
Solid
Fe2O3
Iron(III) Oxide, Rust
[Fe][Fe](=O)(=O)=O
159.69
Glass Bottle
Ferric Oxide, Red, Technical (Iron (III) Oxide)
1565°C
1309-37-1
100 g
MFCD00011008
JEIPFZHSYJVQDO-UHFFFAOYSA-N
trioxodiiron
Technical
5.25
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'