missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Oxide, Red, Technical (Iron (III) Oxide), Reagents
Ferric Oxide, Red, Technical (Iron (III) Oxide), Technical, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C220500025KG
Specifications
| 3414°C | |
| 1565°C | |
| 1309-37-1 | |
| 2.5 kg | |
| MFCD00011008 | |
| JEIPFZHSYJVQDO-UHFFFAOYSA-N | |
| trioxodiiron | |
| Technical | |
| 5.25 |
| Ferric Oxide, Red, Technical (Iron (III) Oxide) | |
| Orange/Red | |
| Solid | |
| Fe2O3 | |
| Iron(III) Oxide, Rust | |
| [Fe][Fe](=O)(=O)=O | |
| 159.69 | |
| Poly Bottle |
Chemical Identifiers
| 1309-37-1 | |
| 159.69 | |
| JEIPFZHSYJVQDO-UHFFFAOYSA-N | |
| trioxodiiron |
| Fe2O3 | |
| MFCD00011008 | |
| Iron(III) Oxide, Rust | |
| [Fe][Fe](=O)(=O)=O |