missing translation for 'onlineSavingsMsg'
Learn More

Ferric Oxide, Red, Technical (Iron (III) Oxide), Reagents

Catalog No. 01337246
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337246 100 g
01337245 2.5 kg
01337244 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337246 Supplier Labchem Inc Supplier No. C2205000100C2
Only null left

Ferric Oxide, Red, Technical (Iron (III) Oxide), Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
Synonym Iron(III) Oxide, Rust
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

Boiling Point 3414°C
Melting Point 1565°C
Color Orange/Red
Physical Form Solid
Quantity 100 g
Grade Technical
Packaging Glass Bottle
Specific Gravity 5.25
Chemical Name or Material Ferric Oxide, Red, Technical (Iron (III) Oxide)