accessibility menu, dialog, popup

UserName

CAS RN 1309-37-1

IUPAC Name: trioxodiiron
Synonyms: trioxodiiron
Molecular Weight (g/mol): 159.69
Molecular Formula: Fe2O3
InChi Key: JEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES: [Fe][Fe](=O)(=O)=O
Physical Form 
  • (2)
  • (18)
  • (5)
  • (4)
  • (4)
Quantity 
  • (3)
  • (5)
  • (2)
  • (1)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
Grade 
  • (3)
  • (2)
  • (5)
  • (7)
  • (1)
Percent Purity 
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (10)
  • (2)
  • (3)
  • (10)
  • (18)
  • (2)
  • (18)
  • (5)
  • (4)
  • (4)
  • (3)
  • (5)
  • (2)
  • (1)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (3)
  • (2)
  • (5)
  • (7)
  • (1)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (3)