missing translation for 'onlineSavingsMsg'
Learn More

Ferric Oxide, Powder, Approx. 95%, Spectrum™ Chemical

$233.50 - $12730.00

Chemical Identifiers

CAS 1309-37-1
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610533
View Documents
Spectrum Chemical Mfg Cor
F1035125GM
125 g Amber Glass Bottle
Each for $233.50
Only null left
 
18610536
View Documents
Spectrum Chemical Mfg Cor
F1035500GM
500 g Amber Glass Bottle
Each for $496.13
Only null left
 
18610535
View Documents
Spectrum Chemical Mfg Cor
F103525KG
2.5 kg Amber Glass Bottle
Each for $1,515.80
Only null left
 
18610534
View Documents
Spectrum Chemical Mfg Cor
F103512KG
12 kg Poly Pail
Each for $4,936.67
Only null left
 
18610537
View Documents
Spectrum Chemical Mfg Cor
F103550KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1309-37-1

Specifications

1309-37-1
125 g
Reagent
1
∼95%
Amber Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.