missing translation for 'onlineSavingsMsg'
Learn More

Ferric Oxide, Powder, Approx. 95%, Spectrum™ Chemical

Catalog No. 18610533
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610533 125 g Amber Glass Bottle
18610536 500 g Amber Glass Bottle
18610535 2.5 kg Amber Glass Bottle
18610534 12 kg Poly Pail
18610537 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610533 Supplier Spectrum Chemical Mfg Cor Supplier No. F1035125GM

Chemical Identifiers

CAS 1309-37-1

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity ∼95%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.