missing translation for 'onlineSavingsMsg'
Learn More

Ferric Oxide, Powder, Approx. 95%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor F1035500GM

 View more versions of this product

Catalog No. 18610536


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1309-37-1
500 g
Reagent
1
∼95%
Amber Glass Bottle

Chemical Identifiers

1309-37-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.