missing translation for 'onlineSavingsMsg'
Learn More

Ferric Nitrate Solution, Stock, 202 g/L in dilute Nitric Acid, Ricca Chemical

Catalog No. 313416
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
313416 500 mL Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 313416 Supplier Ricca Chemical Company Supplier No. 313416
Only null left

Chemical Identifiers

CAS 7697-37-2 , 7782-61-8 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Brown
Grade Laboratory
Packaging Amber Poly Bottle
Solvent or Matrix Dilute Nitric Acid
pH <1
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 202g/L
Chemical Name or Material Ferric Nitrate Solution, Stock Solution