missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solution de nitrate ferrique, pour chlorure, certifiée, LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Ferric Nitrate Solution |
| Nom Remarque | Pour le chlorure |
| Couleur | Vert-Jaune |
| CAS % maximal | 20.2, 77.7, 2.1 |
| Formule linéaire | Fe(NO3)3·9H2O |
| Numéro UN | UN3264 |
| Renseignements sur la solubilité | Soluble dans l’eau |
| Poids de formule | 404.02 |
| Niveau | Certifié |
| Désignation | Test de réussite |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Use only outdoors or in a well-ventilated area. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. Store in a well-ventilated place. Keep container tightly closed. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Température ambiante |