missing translation for 'onlineSavingsMsg'
Learn More

Ferric Nitrate Solution, Stock, 202 g/L in dilute Nitric Acid, Ricca Chemical

Catalog No. 31341
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31341 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31341 Supplier Ricca Chemical Company Supplier No. 31341

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7782-61-8 , 7732-18-5 , 7697-37-2
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
PubChem CID 16211566
IUPAC Name iron(3+) nonahydrate trinitrate
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

CAS Min % 19.0, 79.0, 7.0
CAS Max % 21.0, 83.0, 9.0
Grade APHA/EPA
Packaging Black Poly Bottle
pH <1
Quantity 4 L
Chemical Name or Material Ferric Nitrate Solution, 202g/L in Dilute Nitric Acid