missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride TS, 9% (w/v), Ricca Chemical

Catalog No. 310916
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
310916 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 310916 Supplier Ricca Chemical Company Supplier No. 310916
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10025-77-1 , 7732-18-5
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Color Brown-Orange
Percent Purity 9%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
pH Acidic
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 9% (w/v)
Chemical Name or Material Ferric Chloride, Test Solution