missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride Solution, For Sulfide, 100g in 40mL Water, LabChem™

Catalog No. LC143807
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC143807 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC143807 Supplier LabChem Supplier No. LC143807
Only null left

Chemical Identifiers

CAS 10025-77-1 , 7732-18-5
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Color Amber
CAS Max % 71, 29
Linear Formula FeCl3·6H2O
UN Number UN2582
Concentration 100g in 40mL Water
Solubility Information Soluble in water
Formula Weight 270.3
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride; Iron oxide; Thermal decomposition generates: Corrosive vapors
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Ferric Chloride Solution, For Sulfide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life.
Precautionary Statement GHS P Statement
Do not breathe vapors, mist, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or air): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature