missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride Hexahydrate, ACS, LabChem™

Catalog No. LC142901
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC142901 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC142901 Supplier LabChem Supplier No. LC142901
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 280°C
Melting Point 37°C
Color Brown-Yellow
CAS Max % 100
Physical Form Solid
Quantity 500 g
Linear Formula FeCl3·6H2O
UN Number 3260
Solubility Information Soluble in water
Formula Weight 270.3
Percent Purity 97.0 to 102.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride; iron oxide
Chemical Name or Material Ferric Chloride Hexahydrate
Show More Show Less
Recommended Storage Room Temperature