missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride, as Hexahydrate, Certified, 40.0% (w/v) ±2.0%, LabChem™

Catalog No. LC143704
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC143704 4 L
LC143702 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC143704 Supplier LabChem Supplier No. LC143704

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10025-77-1 , 7732-18-5 , 7647-01-0
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer
PubChem CID 24380  
ChEBI CHEBI:30808
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Color Amber
CAS Max % 40, 54.9, 5.1
Physical Form Liquid
Quantity 4 L
Linear Formula FeCl3·6H2O
UN Number UN3264
Solubility Information Soluble in water
Formula Weight 270.3
Percent Purity 40.0% ±2.0% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.4g/mL
Decomposition Information Hydrogen chloride; Iron oxide; Thermal decomposition generates: Corrosive vapors
Density 1.4g/mL
Chemical Name or Material Ferric Chloride, as Hexahydrate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature