missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Chloride, as Hexahydrate, Certified, 40.0% (w/v) ±2.0%, LabChem™
Specifications
Specifications
| Color | Amber |
| CAS Max % | 40, 54.9, 5.1 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Linear Formula | FeCl3·6H2O |
| UN Number | UN3264 |
| Solubility Information | Soluble in water |
| Formula Weight | 270.3 |
| Percent Purity | 40.0% ±2.0% (w/v) |
| Grade | Certified |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Harmful to aquatic life. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. Avoid release to the environment. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |